thermo-scientific-centrifuges-banner-d
Benchtop Centrifuges
Benchtop Centrifuges
benchtops
Thermo Scientific™ Sorvall™ general purpose centrifuges are designed around you and your applications. With lightweight Fiberlite™ rotors that provide blazingly fast sample processing, these centrifuges can accelerate your research with the flexibility to expand your protocol needs with a complete range of swinging bucket, fixed angle and carbon fiber rotors.
General Purpose Centrifuges
TX-1000
More Samples
  • 4L capacity in a traditional 3L footprint
  • Maximized capacity up to 40 x 50mL conical tubes, 6 x 250mL and 4 x 1000mL bottles on the bench
  • Thermo Scientific™ Auto-Lock™ rotor exchange for system versatility
More Speed
High RPM/RCF with both swinging bucket and fixed angle configurations, including Thermo Scientific™ Fiberlite™ carbon fiber rotors.
More Savings
Limited-time offers on packages designed around you and your applications — Thermo Scientific Sorvall general-purpose centrifuges, rotors and accessories.
  • Five-year centrifuge warranty (no registration necessary)
  • One-handed certified Thermo Scientific™ ClickSeal™ biocontainment lids at no additional cost
  • 15-year Fiberlite rotor warranty
We'll get you off the bench and on to answers faster, any way you spin it.
  • Thermo Scientific Sorvall Legend XT/XF Centrifuge Series
    More capacity, performance (up to 25,830 x g) and ergonomics in a compact footprint. Thermo Scientific™ Sorvall™ Legend™ XF floor-standing configuration is also available to save valuable lab space.
    Learn More
  • Thermo Scientific Sorvall Legend X1 Centrifuge Series
    Compact 1.6L capacity supports exceptional throughput with a broad rotor range for versatility and high-performance processing.
    Learn More
Small Benchtop Centrifuges
Quick, reliable and safe separations with Thermo Scientific small benchtop centrifuges provide versatility for numerous applications and improve lab efficiency in a compact footprint to maximize workspace.
  • NEW! Thermo Scientific Sorvall ST 8 Centrifuge
    Maximizes capacity with 8 x 50 mL conical tubes, microplates and microtubes, all in a compact design to accommodate tight lab spaces across both clinical and research applications.
    Learn More
  • Thermo Scientific™ Medilite™ Centrifuge
    An excellent choice for the budget-minded lab.
    Learn More
  • Thermo Scientific Sorvall Stratos Centrifuge Series
    Versatile, refrigerated high-speed centrifuge achieves up to 50,000 x g to process samples with exceptional performance, flexibility and reliability.
    Learn More
Microcentrifuges
Experience a new separation experience with Thermo Scientific microcentrifuges that combine power, versatility and convenience in a safe, compact, easy-to-use lab instrument.
  • Thermo Scientific Sorvall Legend Micro 17 and 21 Microcentrifuge Series
    From the intuitive controls, to the innovative ClickSeal™ biocontainment lid, the Sorvall Legend Micro 17 and 21 microcentrifuges deliver the perfect blend of capabilities to support your micro-volume protocols.
    Learn More
Floor Centrifuges
Floor Model Centrifuges
FloorModels
Ensure consistent results year after year, as well as the operational longevity of your system, with:
  • Innovative design
  • High throughput and speed
  • Operator, sample and system safety
These products are not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for assistance.
Superspeed Centrifuges
Meet evolving application needs, from academic research to production facilities, with up to 100,605 x g, and up to a 6L capacity.
  • NEW! Thermo Scientific™ Sorvall™ LYNX Superspeed Centrifuge Series
    Simplify productivity and safety with powerful Thermo Scientific technologies:
    • Push-button security and application flexibility with Auto-Lock™ rotor exchange
    • Immediate rotor detection and programming with Auto-ID instant rotor identification
    • Improved ergonomics and performance with Fiberlite™ carbon fiber rotors
    • One-handed sample protection certified by Public Health England, Porton Down, UK with ClickSeal™ biocontainment lids
    Learn More
Ultracentrifuges
Powerful separation technology in a quiet, compact design.
  • Thermo Scientific™ Sorvall™ WX Ultracentrifuges
    Meet your high throughput needs with Thermo Scientific Fiberlite rotors to maximize centrifuge performance.
    Learn More
  • Thermo Scientific™ Sorvall™ MX Plus Micro-ultracentrifuge
    Available in two speeds, with a full range of rotors for small volume processing of 0.2 mL to 30 mL tubes.
    Learn More
  • Thermo Scientific™ Sorvall™ MTX 150 Ultracentrifuge
    The power of ultracentrifugation, up to 1,048,000 x g in a small, quiet and fast benchtop micro-ultracentrifuge.
    Learn More
Large Capacity Centrifuges
  • Thermo Scientific™ Sorvall™ RC12BP Plus Large Capacity Centrifuge
    An integrated system for high-throughput, large volume blood-banking and bioprocessing applications.
    Learn More
  • Thermo Scientific™ Sorvall™ RC3BP Plus Large Capacity Centrifuge
    Combines dependable performance and ease-of-use with advanced functionality that maximizes reproducibility and throughput.
    Learn More
Special Offers
Resources
More
Other Thermo Scientific Resources