Learn More
JP Selecta™ Alcohol Burners
Suitable for general laboratory purpose.JP Selecta™ Alcohol Burner is chrome plated.
8.20€ - 34.80€
Specifications
Product Type Alcohol Burner
Weight (Metric) 0.01 kg
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
12027344
View Documents
JP Selecta™
1001434
85 mL
34.80€
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
12062944
View Documents
JP Selecta™
1001256
120 mL
8.20€
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Has lid and chain
  • Has screwcap jar, chrome plated top with wick height control
Specifications
Specifications
Alcohol Burner
0.01 kg