Learn More
ANXA9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 15416-1-AP-20UL
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16874574
  • 145.00 € / 20 Mikroliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Entdecken Sie weitere Sonderangebote
Beschreibung
Beschreibung
The annexins include ANXA9 coded protein, which are a family of calcium-dependent phospholipid-binding proteins. Members of the annexin family contain 4 internal repeat domains, each of which includes a type II calcium-binding site. The calcium-binding sites are required for annexins to aggregate and cooperatively bind anionic phospholipids and extracellular matrix proteins. Annexin A9 is a divergent member of the annexin protein family in which all four homologous type II calcium-binding sites in the conserved tetrad core contain amino acid substitutions that ablate their function. However, structural analysis suggests that the conserved putative ion channel formed by the tetrad core is intact.
Spezifikation
Spezifikation
ANXA9
Polyclonal
Unconjugated
ANXA9
Annexin 31, Annexin 9, annexin A9, Annexin XXXI, ANX31, ANXA9, Pemphaxin
Rabbit
Antigen Affinity Chromatography
RUO
71790, 8416
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O76027, Q9JHQ0
Anxa9
ANXA9 Fusion Protein Ag7702
20 μL
Primary
Mouse, Human