Learn More
Karl Hecht Assistent™ Glass Beads
Made of clear glass. Karl Hecht™ Assistent™ Glass Beads can be used for packing distillation columns.
Brand: Karl Hecht HEC-1401/4
This item is not returnable.
View return policy
Description
- Solid
- Spherical
- Clear glass
- Packs of 1kg
Specifications
Beads | |
Glass | |
4 mm | |
Distillation Columns |