Learn More
Telstar™ AV-30/70 Vertical Laminar Flow Bench
The AV-30/70 Vertical Laminar Flow Bench, with 2 HEPA filters, 70% down flow and 30% exhaust, provides underpressure conditions in the working area, therefore giving a good protection level to the product, to the operator as well as environment.
Brand: Telstar™ 31401
This item is not returnable.
View return policy
Description
- Supplied with working area CLASS 10 as defined by U.S. Fed. Std. 209 a B.S. 5295
- Specially designed for handling of non-pathogenic biological samples, cell and tissue cultures, controls in microbiology, pharmaceutical products preparation, etc.
- Also suitable for work with electronics and optics
- Cabinets made of stove-enamel coated steel
- Modern and accurate design makes it possible to change the filters from inside the cabinet, having reduced external dimensions with suitable working area
- Polished stainless steel working surface divided in three parts, easily removable for sterilizing
- Removable side walls and front window wiht gas springs, both made of UV resistant tempered glass
- Materials and construction according to international standards, with high quality components and easy service
- Digital control board with LCD display including selection of air speed, button for UV irradiation, alarm LED, time counter for the fan and more
- ON/OFF selector
- Electrical power socket inside of the cabinet
Specifications
Stove Enamel | |
Stove Enamel-coated Steel, Polished Stainless Steel (Work Surface) | |
Handling of non-pathogenic biological samples, cell and tissue cultures, controls in microbiology, pharmaceutical products preparation, electronics, optics, etc. | |
50 Hz, Single Phase | |
1.7 kW | |
150 kg |
HEPA | |
845 x 1260 x 1445 mm | |
700 mm | |
>800 lx | |
220 V |