Learn More
Sampling Systems™ Blue SteriWare™ Liquid Funnels
Remove the risk of cross contamination and improve your quality control practices by dispensing liquids with blue SteriWare™ Liquid Funnels. Manufactured in our cleanroom and available sterile, these funnels are ideal for use in a wide range of applications in the food industry.
60.45 € - 147.00 €
Spezifikation
Oberer Durchmesser (metrisch) | 100 mm |
---|---|
Material | Polypropylen |
Farbe | Blau |
Produktcode | Marke | Sterilität | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Sterilität | Preis | Menge & Verfügbarkeit | |||||
17574656
![]() |
Sampling Systems™
8370B-100 |
Unsteril |
60.45 €
50 Stück |
Verfügbar ab: 23-07-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
Sie haben bereits ein Konto? Melden Sie sich an um den Preis und die Verfügbarkeit zu sehen. Neu hier? Registrieren Sie sich noch heute bei uns! |
||||
17584656
![]() |
Sampling Systems™
8370B-100S |
Steril |
147.00 €
50 Stück |
Verfügbar ab: 23-07-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
Sie haben bereits ein Konto? Melden Sie sich an um den Preis und die Verfügbarkeit zu sehen. Neu hier? Registrieren Sie sich noch heute bei uns! |
||||
Beschreibung
- Our range of single use, disposable liquid funnels are ideal for use in laboratories and manufacturing areas.
- These funnels are made from food grade polymers and are designed to be used straight from the bag.
- The bright blue color means they can be easily seen if they drop in or left in the material by mistake.
- SteriWare™ Funnels are available sterile and non-sterile.
- SteriWare™ is the largest range of single-use equipment for sampling, laboratory and production work on the market.
- The whole range is made in a cleanroom and comes with class-leading pharmaceutical documentation.
- Simply unwrap the funnel and use.
- The funnels come with a side tab so they are easy to hold.
- The individually bagged funnels are made from PP that is FDA/EC compliant.
- The material used is also certified BSE/TSE free. The sterile funnels conform to SAL 10-6
- The funnels come with a batch-specific certificate that includes the date of manufacture, expiry date and irradiation certificate (for sterile funnels only).
Spezifikation
100 mm | |
Polypropylen | |
Blau |