Learn More
CD117 Rat anti-Mouse, APC, Clone: 2B8, Proteintech
Rat Monoclonal Antibody
Marke: Proteintech APC-65054-100UG
Beschreibung
The CD117 antibody from Proteintech is a rat monoclonal antibody generated with mouse il-3-dependent bone marrow mast cells of mouse CD117. This antibody recognizes mouse antigen. The CD117 antibody has been validated for the following applications: FC analysis. This antibody is apc.
Spezifikation
CD117 | |
Monoclonal | |
0.1 mg/mL | |
Rat | |
Primary | |
Mouse |
Flow Cytometry | |
2B8 | |
APC | |
100 μg | |
16590 | |
IgG2b |