Learn More
Thermo Scientific™ Temperature Control Connectivity Accessories
Maximize the efficiency of your temperature control device with Thermo Scientific™ Temperature Control Connectivity and monitoring accessories.
Brand: Thermo Scientific™ 1600033
1058.74 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Maximize the efficiency of your temperature control device with Thermo Scientific™ Temperature Control Connectivity and monitoring accessories.
Connect your Thermo Scientific temperature control solution with your application.- Available for temperature control devices equipped with communications
- Designed to work with your Thermo Scientific recirculating chiller or bath equipped with communications
- Available in popular configurations that match your device’s connections and interface with your process
- Take the guess work out of finding parts to match your recirculating chiller or bath
- Provides remote control of Thermo Scientific units equipped with an RS-232 communications interface
- Choose upper and lower temperature alarm limits and monitor system status with an alarm
- Record your results on a user selectable graph
- Select from easy–to–use product icons
- Create graphs and charts
- Easily configure ramping functions over time
- View pop-up alarm windows which display the error condition
- View a virtual controller screen
- PC compatible only
Specifications
Interface Cable,USB, 1.8m | |
USB | |
Thermo Scientific™ Refrigerated and Heated Bath Circulators |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.