Learn More
Reichert™ Cuvets for Reichert Unistat™ Bilirubinometer
For use with Reichert UNISTAT Bilirubinometer (cat. no. AO10310C)
Brand: Reichert™ 13103430
657.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Low-Level Check Cuvette
- For use with Reichert Unistat Bilirubinometer
- Capacity: 20μL
Specifications
Low to Level Check Cuvette | |
Undiluted Serum | |
Reichert Unistat™ Bilirubinometer | |
20 μL |