Learn More
JP Selecta™ Drying and Sterilization Oven
Multipurpose, fast response and recuperation of temperature.
Brand: JP Selecta™ 2000201
This item is not returnable.
View return policy
Description
- TFT touch screen
- Inner chamber made of AISI 304 stainless steel
- Pre-mixing chamber made of AISI 304 stainless steel
- Homogeneously distributed shielded heating elements with complete air circulation
- Low external temperature due to excellent thermal insulation
- Flexible silicon door gasket around the entrance of chamber
- Door seal due to floating inner door that adjusts and absorbs thermal expansion
- Independent insulated control box
- Epoxy coated outer case
- Ventilator with adjustable outlet (access at the back of the unit)
- Adjustable height positions for guides and shelves
- Toughened double safety glass door for viewing
Specifications
68 cm | |
150 L | |
Drying and sterilization | |
70 cm | |
Oven | |
50 cm | |
Stainless Steel (AISI 304) | |
76 kg | |
60 cm |
50 cm | |
50 cm | |
Drying and sterilization | |
2000201 | |
5°C to 250°C | |
Analogue thermometer | |
2150 w | |
8 | |
95 cm |