Learn More
Tocris Bioscience™ ERK5-IN-1
Potent and selective ERK5 inhibitor
Brand: Tocris Bioscience™ 5393/50
This item is not returnable.
View return policy
Description
ERK5-IN-1 is a potent and selective ERK5 inhibitor; inhibits EGFR-induced ERK5 autophosphorylation (EC50 = 90 nM) and ERK5 enzymatic activity (IC50 = 162 nM). Exhibits at least 30-fold selectivity for ERK5 over LRRK2 in a cell based assay. Also selective for ERK5 over a panel of other kinases. Orally bioavailable.
Specifications
>98% | |
638.8 | |
50 mg |
ERK5-IN-1 | |
ERK5-IN-1 |