Learn More
Hettich Lab Technology™ Fixed Angle Rotor
Fixed Angle Rotor has a relatively low drag for high-speed centrifugation and oblique angling to speed up sedimentation.
Brand:  Hettich Lab Technology™ 1613
348.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10191372
  • 382.00€ / Each
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description
Description
  • Used primarily in the higher speed range.
  • Relatively low drag.
  • Oblique angling of tubes shortens the overall distance that particles must travel, thereby speeding up sedimentation.
Specifications
Specifications
Fixed Angle Rotor
4,146 x g
125 mm
Universal 320, Rotofix 32A
12
12 x 1.1 to 15 mL
6,000 rpm
17 mm
35°
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products