Learn More
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11941319
|
Afora™
141 |
2000 mL |
522.00€
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||||
12047673
|
Afora™
V57439 |
202.00€
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
12080565
|
Afora™
V53420 |
500 mL |
318.00€
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
|||||
Specifications
Reactor |