Learn More
GAS1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 17903-1-AP-20UL
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16877904
  • 145.00 € / 20 Mikroliter
Please to purchase this item. Need a web account? Register with us today!
Entdecken Sie weitere Sonderangebote
Beschreibung
Beschreibung
Growth arrest-specific 1 plays a role in growth suppression. GAS1 blocks entry to S phase and prevents cycling of normal and transformed cells. Gas1 is a putative tumor suppressor gene.
Spezifikation
Spezifikation
GAS1
Polyclonal
Unconjugated
GAS1
GAS 1, GAS1, growth arrest 1
Rabbit
Antigen Affinity Chromatography
RUO
2619
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P54826
GAS1
GAS1 Fusion Protein Ag12275