Learn More
IL-9 Mouse anti-Human, CoraLite 488, Clone: 1F3A11, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL488-66144-100UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IL-9 Monoclonal antibody apecifically detetects IL-9 antigen in Human Samples. It is validated for Immunofluorescence.Spezifikation
IL-9 | |
Monoclonal | |
1000 μg/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
IL9 | |
IL-9 fusion protein Ag21435 | |
100 μL | |
3578 | |
Store at -20°C. Avoid exposure to light. | |
IgG2b |
Immunofluorescence | |
1F3A11 | |
CoraLite 488 | |
BC066284 | |
Mouse | |
Protein A purification | |
Primary | |
Human | |