Learn More
VACUUBRAND™ KF DN 10mm Screw Flange With G 3/8 in. Internal Thread
Brand:  VACUUBRAND™ 20643813
Product Code. 10127434
  • 705.00 DKK / Each
Only null left
Estimated Shipment: 01-09-2025
Add to basket
Specifications
1 Each
KF DN 10 mm
Screw Flange