Learn More
Liebherr™ KTS Series Table-top Refrigerator/Freezer
Brand: Liebherr™ KTS 127
This item is not returnable.
View return policy
Specifications
112 L | |
2 | |
R600A | |
White | |
62 cm | |
50 cm | |
33.4 kg |
3°C to 8°C | |
Automatic | |
CE, ISO 9001 | |
Static | |
82 cm | |
230 V |