Learn More
Brand™ LDPE Wash Bottles without Valve
Manufactured from high-quality, flexible LDPE. BRAND™ LDPE Wash Bottles without Valve include an integrated wash stream and sealing cone closure. Large opening provides convenient filling. Label designates contents and includes general safety statements in English (with CAS numbers and NFPA codes) and German (with CAS numbers).
29.50€ - 44.50€
Specifications
Material | LDPE |
---|---|
Mouth | Wide |
Product Type | Wash Bottle |
Closure Type | Sealing Cone |
Product Code | Brand | Capacity (Metric) | Chemical Label | Closure Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Chemical Label | Closure Color | Price | Quantity & Availability | |||
10673582
|
Brand™
144079 |
250 mL | Distilled Water | White | |||||
10355421
|
Brand™
144084 |
500 mL | Distilled Water | White | |||||
10528211
|
Brand™
144089 |
500 mL | Neutral | Yellow | |||||
10693772
|
Brand™
144086 |
500 mL | Methanol | Green | |||||
10548021
|
Brand™
144087 |
500 mL | Isopropanol | Blue | |||||
10739391
|
Brand™
144085 |
500 mL | Acetone | Red | |||||
10603782
|
Brand™
144088 |
500 mL | Ethanol | Orange | |||||
Description
- Manufactured from flexible LDPE
- Wide-mouth.
- High-quality wash bottles with integrated wash stream and sealing cone closure.
- Large opening for convenient filling.
- Label with designation of contents and general safety statements in English (with CAS numbers and NFPA codes) and German (with CAS numbers).
Specifications
LDPE | |
Wide | |
Wash Bottle | |
Sealing Cone |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.