Learn More
Lonza™ BioWhittaker™ ProNSO™ CD Media
Chemically defined, protein-free medium designed to meet a growing demand for optimized NS0 formulations
Spezifikation
Form Flüssig
Produktlinie ProNS0™
Mehr Spezifikationen ansehen
Beschreibung
Beschreibung
These NS0 products provides maximum protein productivity. Two distinct proprietary media formulations are offered to cover a broad range of nutritional needs; try each to determine which one provides the best yield for your application.
  • Further maximize protein production by titration with ProNS0 Lipid CD Supplement
  • Formulations contain HEPES and Pluronic, without L-glutamine, phenol red, or cholesterol
  • Product purification is simplified
  • Lot-to-lot consistency ensures dependable performance
  • Non-animal origin components reduce regulatory burdens
Spezifikation
Spezifikation
Flüssig
ProNS0™