Learn More
Ansell™ Microchem™ 3000 PAPR Encapsulated Coveralls
Product Code. 15556584 Shop All Ansell Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Provides head and body protection from hazardous substances. MICROCHEM™ 3000 PAPR Encapsulated Coveralls feature a soft and flexible 3-layer fabric, strong ultrasonically welded seams and an effective chemical barrier against most inorganic chemicals.
  • Fully encapsulated coverall, for use in combination with Scott Proflow PAPR fan unit, breathing hose and filters.
  • Double elasticated cuffs, attached socks with boot over-flap.
  • Emergency rip cord attached to left chest shoulder and left back shoulder as worn
  • Multi-layer barrier fabric effective against numerous chemicals
  • Highly visible bright yellow for improved worker safety
  • Lightweight yet durable
  • Anti-static according to EN 1149-5
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Type 3/4 EN14605, Type 5 EN ISO 13982-1, EN14126, EN1073-2, EN 12941, EN1149-5
Color Yellow
Hood Type Fully Encapsulated
Material Microporous Polypropylene Laminate
Size Large
Ankle Style Attached socks, Boot overflaps
Back Style Accommodates Scott Proflow 2SC/EX PAPR fan unit, breathing hoses, and filters
Collar Type Air permeable SMS
Cuff Style Double, Elastic
Description Emergency rip cord
For Use With (Equipment) Scott Proflow 2SC/EX PAPR fan unit, breathing hoses, and filters
No. of Pockets None
Product Type Encapsulated coveralls
Seam Type Stitched, Taped
Style Includes Emergency rip cord