Learn More
Comecta™ HEROLAB Polypropylene Centrifuge Tubes
Plastic centrifuge tubes with lids are made from translucent polypropylene, autoclavable.
39.00€ - 43.68€
Specifications
Product Type | Centrifuge Tube |
---|---|
Autoclavable | Autoclavable |
Material | Polypropylene |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12007684
|
Comecta™
5753411 |
250 mL |
43.68€
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
|||||
12097674
|
Comecta™
5753243 |
80 mL |
39.00€
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
|||||
Description
- Available in 10 mL to 1000 mL capacity
- Autoclavable
Specifications
Centrifuge Tube | |
Autoclavable | |
Polypropylene |