Learn More
Cytiva Polypropylene Plunger
46.00€ - 53.00€
Specifications
Material | Polypropylene (Reinforced Glassfibre) |
---|---|
Type | Plunger |
Max. Temperature (Metric) | 60°C |
Quantity | 1 Pack |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10630571
|
Cytiva
18-1031-80 |
XK 16 and AK 16 Columns |
46.00€
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
|||||
10327889
|
Cytiva
18-1031-81 |
XK 26, AK 26, and AK 16 |
53.00€
Each |
In Stock
Log in to see stock. |
|||||
Description
Specifications
Polypropylene (Reinforced Glassfibre) | |
Plunger | |
60°C | |
1 Pack |