Learn More
Technical Service Consultants™ Polystyrene L-shaped Spreader
Use Technical Service Consultants™ Polystyrene L-shaped Spreader for surface spreading of liquid or culture samples uniformly over the surface of solid media. Shaped like a hockey stick, it is recommended for single use practices or sterile techniques.
1025.00 DKK - 1415.00 DKK
Specifications
Material | Polystyrene |
---|---|
Product Type | Spreader |
Sterility | Sterile |
Shape | L-Shape |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12749885
|
Technical Service Consultants™
TS/30-B |
500 Pk. |
1025.00 DKK
Pack of 500 |
Estimated Shipment: 03-10-2025
Log in to see stock. |
|||||
12739885
|
Technical Service Consultants™
TS/30-A |
1000/Pk. |
1415.00 DKK
Pack of 1000 |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||||
Description
- Polystyrene
- Three year shelf life
- Ideal for bacterial counts
- L shaped plastic (firm) peel pouch
- Hockey Stick Shape
Specifications
Polystyrene | |
Spreader | |
Sterile | |
L-Shape |