Learn More
PRKAR1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 17991-1-AP-150UL
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16868014
  • 410.00 € / 150 Mikroliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Entdecken Sie weitere Sonderangebote
Beschreibung
Beschreibung
The second messenger cyclic AMP (cAMP) mediates diverse cellular responses to external signals such as proliferation, ion transport, regulation of metabolism and gene transcription by activation of the cAMP-dependent protein kinase (cAPK or PKA). Activation of PKA occurs when cAMP binds to the two regulatory subunits of the tetrameric PKA holoenzyme resulting in release of active catalytic subunits. Three catalytic (C) subunits have been identified that each represent specific gene products. Activation of transcription upon elevation of cAMP levels results from translocation of PKA to the nucleus where it phosphorylates the transcrip- tion factor cAMP response element binding protein (CREB) on serine 133 which in turn leads to TFIIB binding to TATA-box-binding protein TBP1, thus linking phospho-CREB to the pol II transcription initiation complex.
Spezifikation
Spezifikation
PRKAR1B
Polyclonal
Unconjugated
PRKAR1B
PRKAR1, PRKAR1B
Rabbit
Antigen Affinity Chromatography
RUO
19085, 25521, 5575
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P12849, P31321, P81377
Prkar1b
PRKAR1B Fusion Protein Ag12426
150 μL
Primary
Human, Mouse, Rat