Learn More
Afora™ Reactor Cover
Use the Afora™ Reactor Cover in conjunction with this product.
348.00€ - 672.00€
Product Code | Brand | Diameter (Metric) Outer | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Includes | Price | Quantity & Availability | ||||
11996358
|
Afora™
108 |
75 mm | Center arm: 34/35; Side arms: N/A | N/A | |||||
11981199
|
Afora™
111/E |
100 mm | Center arm: 29/32; Side arms: 14/24 (0°), 14/23 (5°), 14/24 (10°), 29/32 (15°) | N/A | |||||
12079417
|
Afora™
113/E |
100 mm | Center arm: 29/32; Side arms: 14/24 (0°), 14/23 (5°), 29/32 (10°), 29/32 (15°) | N/A | |||||
11901129
|
Afora™
110/E |
100 mm | Center arm: 19/26; Side arms: 14/24 (0°), 14/23 (5°), 14/24 (10°), 29/32 (15°) | N/A | |||||
12033908
|
Afora™
113 |
100 mm | Center arm: 29/32; Side arms: 14/23 (0°), 14/23 (5°), 29/32 (10°), 29/32 (15°) | N/A | |||||
Description
- Reactor accessory
Specifications
Reactor Cover |