Learn More
Witeg™ Suction Bulb for Pipettes
A vacuum source for filling reagents and solutions.
Brand:  Witeg™ 3968001
Product Code. 15741928
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • Green in color
  • 1 mL capacity
  • Provided 10 per pack
Specifications
Specifications
Green
1 mL
Filling/transfer
Pipettes