Learn More
Thermo Scientific™ SUN-SRi™ Dry Vials™
Nitrogen-purged vials used for low-level water determination in the tobacco industry
41.00€
Product Code | Brand | dimColor | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | dimColor | Quantity | Price | Quantity & Availability | ||||
12054598
|
Thermo Scientific™
2001/14 |
N/A | |||||||
Description
- Standard-opening, 1.5mL, 12 × 32mm vials for use in FTC regulation Volume 32 #147 and Health Canada Official Method T-115 to determine the amount of tar, nicotine, and carbon monoxide in tobacco smoke
- Nitrogen purged to provide true total particulate matter readings free of contaminating water/water vapor from vial surfaces
- Includes aluminum seal closures with PTFE/rubber septa
- Available in Clear, Type 1 Class A or Amber, Type 1 Class B borosilicate glass