Learn More
Syntaxin 16 Mouse anti-Human, CoraLite 594, Clone: 4A9D8, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL594-66775-100UL
Beschreibung
The Syntaxin 16 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Syntaxin 16. This antibody recognizes human antigen. The Syntaxin 16 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Spezifikation
Syntaxin 16 | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human |
Immunofluorescence | |
4A9D8 | |
CoraLite 594 | |
100 μL | |
8675 | |
IgG1 |