Learn More
Rogo Sampaic™ Transparent Glass Coverslips
Glass coverslips are transparent and don't leave fingerprints.
Spezifikation
Form | Square |
---|---|
Material | Kalknatronglas |
Dicke (metrisch) | Nr. 1 (0,13 bis 0,16 mm) |
Beschreibung | Glass Coverslip |
Menge | 100 |
Beschreibung
Glass coverslips are transparent and don't leave fingerprints.
- Manufactured in soda glass
- Uniform thickness of 0.13 to 0.16 mm
Spezifikation
Square | |
Nr. 1 (0,13 bis 0,16 mm) | |
100 |
Kalknatronglas | |
Glass Coverslip |