Learn More
Greiner Bio-One Ultra Low Profile Polystyrene Microplate Lids
GREENER_CHOICE
Used for high-format low profile microplates
112.79€
Specifications
Color Clear
Material Polystyrene
Condensation Rings No
No. per Case 100
No. per Pack 5
View More Specs
Products 1
Product Code Brand Sterility Price Quantity & Availability  
Product Code Brand Sterility Price Quantity & Availability  
10398623
GREENER_CHOICE
View Documents
Greiner Bio-One
691101
Non-sterile
112.79€
Pack of 100
Only null left
Estimated Shipment: 25-07-2025
Add to basket
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic, non-cytotoxic
  • Offer protection against contamination and evaporation during sample storage and cell cultivation
  • Compatible with a range of different microplates
Specifications
Specifications
Clear
No
5
Polystyrene
100
A variety of microplates, particularly well suited for high-format, low-profile microplates
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.