Learn More
Afora™ Volumetric Tube
29.00€ - 99.00€
Specifications
Product Type | Volumetric Tube |
---|---|
Quantity | 1 Pack |
Packaging Quantity | 1 Pack |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12080765
|
Afora™
V53623 |
300 mL |
99.00€
Each |
Estimated Shipment: 11-07-2025
Log in to see stock. |
|||||
12020555
|
Afora™
V53402 |
50 mL |
29.00€
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
|||||
Specifications
Volumetric Tube | |
1 Pack | |
1 Pack |