missing translation for 'onlineSavingsMsg'
Learn More

Occludin Rabbit anti-Human, Mouse, Rat, CoraLite 488, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech CL488-13409-20UL

 View more versions of this product

Product Code. 17701259

  • 145.00 EUR / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Occludin Polyclonal antibody specifically detects Occludin in Human, Mouse, Rat samples. It is validated for IF
Specifications

Specifications

Occludin
Polyclonal
CoraLite 488
BC006494
Antigen affinity purification
Primary
Human, Mouse, Rat
Antibody
IgG
Immunofluorescence
1000 μg/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Rabbit
20 μL
4950
Store at -20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.