Learn More
TMEM100 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25581-1-AP-20UL
This item is not returnable.
View return policy
Description
The TMEM100 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human TMEM100. This antibody recognizes human, mouse antigen. The TMEM100 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
TMEM100 | |
Polyclonal | |
Unconjugated | |
20 μL | |
55273 | |
IgG |
Immunohistochemistry, ELISA | |
467 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |